Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 300 Å, 3.5 μm
Supplier:  Waters Corp 186003610
Encompass_Preferred
  • Optimized and QC tested for peptide separations to ensure method stability.
  • Features wide-pore (300Å) trifunctionally bonded BEH particles.
  • Offers the widest usable pH range (1 to 12).
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed.
  • Manufactured in a cGMP, ISO 9002 certified plant using ultra-pure reagents.
  • Designed to provide excellent peak shape and high efficiency.
  • Incorporates BEH technology and uses synthetic particles for high quality and consistent performance.
Catalog No. 50-781-886