Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 300Å, 3.5 μm, 4.6 mm x 100 mm
Catalog No. 50781888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH300 C18 PST columns are optimized and QC tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, offering a wide pH range (1-12), superior low pH stability, and ultra-low column bleed. This ensures reliable batch-to-batch separations for proteomics, protein characterization, and peptide synthesis.
  • C18 ligand with well-characterized, cutting-edge bonding techniques ensures predictable behavior
  • Robust particle structure and bonding chemistry ensure stability across a wide pH range and high temperatures
  • Packing materials produced in a cGMP, ISO 9002 certified facility using ultra-pure reagent
  • Includes a Performance Test Chromatogram and Certification of Acceptance
Catalog No. 50-781-888 Supplier Waters Corp Supplier No. 186003612
XBridge BEH300 C18 PST columns are optimized and QC tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, offering a wide pH range (1-12), superior low pH stability, and ultra-low column bleed. This ensures reliable batch-to-batch separations for proteomics, protein characterization, and peptide synthesis.
  • C18 ligand with well-characterized, cutting-edge bonding techniques ensures predictable behavior
  • Robust particle structure and bonding chemistry ensure stability across a wide pH range and high temperatures
  • Packing materials produced in a cGMP, ISO 9002 certified facility using ultra-pure reagent
  • Includes a Performance Test Chromatogram and Certification of Acceptance