Learn More
Waters Corp Waters C18 Column, 300Å, 5 μm, 4.6 mm x 150 mm
Catalog No. 50781900 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are optimized and QC tested for peptide separations, featuring wide-pore (300Å) BEH particles, wide pH range (1 to 12), and superior low pH stability, making them suitable for proteomics and protein characterization.
  • Optimized and QC tested for peptide separations, featuring wide-pore (300Å) BEH particles with a 5 μm particle size.
  • Offers a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Suitable for proteomics, protein characterization, and peptide synthesis applications.
  • Provides high quality and consistent performance with BEH technology, withstanding pressures up to 6000 psi (415 Bar).
  • Manufactured in a cGMP, ISO 9002 accredited facility, ensuring good peak shape, high efficiency, and stability.
  • Each column is individually tested and includes a performance test chromatogram and certificate of acceptance.
Catalog No. 50-781-900 Supplier Waters Corp Supplier No. 186003624
These columns are optimized and QC tested for peptide separations, featuring wide-pore (300Å) BEH particles, wide pH range (1 to 12), and superior low pH stability, making them suitable for proteomics and protein characterization.
  • Optimized and QC tested for peptide separations, featuring wide-pore (300Å) BEH particles with a 5 μm particle size.
  • Offers a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Suitable for proteomics, protein characterization, and peptide synthesis applications.
  • Provides high quality and consistent performance with BEH technology, withstanding pressures up to 6000 psi (415 Bar).
  • Manufactured in a cGMP, ISO 9002 accredited facility, ensuring good peak shape, high efficiency, and stability.
  • Each column is individually tested and includes a performance test chromatogram and certificate of acceptance.