Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Peptide BEH C18 Column, 300Å, 5 μm

Catalog No. 50781892 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized for peptide separations. They feature wide-pore (300Å) trifunctionally bonded BEH particles, offering a broad pH range (1-12), superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations.
  • Features wide-pore (300Å) BEH particles with trifunctional bonding, ensuring ultra-low column bleed.
  • Offers a usable pH range of 1 to 12 with superior low pH stability.
  • C18 chemistry and Reversed Phase separation mode, ideal for proteomics.
  • Hybrid, spherical particles, 5 μm particle size.
  • Maximum pressure capacity: 6000 psi (415 Bar).
  • Dimensions: 1 mm inner diameter, 100 mm length.
  • 18% carbon load; compatible with HPLC systems.

Catalog No. 50-781-892 Supplier Waters Corp Supplier No. 186003616
May include imposed supplier surcharges.
Add to Cart
Add to Cart

These XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized for peptide separations. They feature wide-pore (300Å) trifunctionally bonded BEH particles, offering a broad pH range (1-12), superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations.
  • Features wide-pore (300Å) BEH particles with trifunctional bonding, ensuring ultra-low column bleed.
  • Offers a usable pH range of 1 to 12 with superior low pH stability.
  • C18 chemistry and Reversed Phase separation mode, ideal for proteomics.
  • Hybrid, spherical particles, 5 μm particle size.
  • Maximum pressure capacity: 6000 psi (415 Bar).
  • Dimensions: 1 mm inner diameter, 100 mm length.
  • 18% carbon load; compatible with HPLC systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.