Learn More
Waters Corp Waters XBridge Peptide C18 Prep Column, 300 Å, 5 μm
Supplier: Waters Corp 186003630
These C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded particles offer a wide usable pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.
- Optimized and QC tested for peptide separations.
- C18 chemistry, reversed phase, with 5 μm particles and 300 Å pores.
- Wide pH range (1 to 12) and stable at high temperatures.
- Maximum pressure 6000 psi (415 bar); low silanol activity.
- Molecular weight range: 1,000 to 15,000.
- OBD Prep Column: 19 mm ID × 50 mm length.
- Superior peak capacity and shape in trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents for peptides and small proteins.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.