Learn More
Waters Corp Waters Corporation XBridge Peptide BEH C18 Column, 300Å, 5 μm, 4.6 mm x 250 mm
Supplier:  Waters Corp 186003625
Encompass_Preferred
These BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. Featuring a wide-pore (300Å) trifunctionally bonded BEH particle, they offer a wide usable pH range (pH 1 to 12), superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Widest usable pH range (pH 1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.
  • Ensures consistent batch-to-batch synthetic peptide and protein digest separations.
  • Every column tested, with a Performance Test Chromatogram and Certification of Acceptance included.
Catalog No. 50-781-901