Learn More
Waters Corp Waters Corporation XBridge Peptide BEH C18 Column, 300 Å, 5 μm, 2.1 mm × 100 mm
Supplier:  Waters Corp 186003619
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Widest usable pH range (1 to 12) with superior low pH stability.
  • Ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.
  • Utilizes cutting-edge C18 ligand bonding techniques.
  • Ensures consistent synthetic peptide and protein digest separations.
  • Developed in a cGMP and ISO 9002 recognized facility.
  • Tested and includes Performance Test Chromatogram and Certificate of Acceptance.
Catalog No. 50-781-895