Learn More
Waters Corp Waters Column, XBridge Peptide BEH C18, 300Å, 5 μm, 2.1 mm x 250 mm
Supplier:  Waters Corp 186003621
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. These columns feature wide-pore (300Å) trifunctionally bonded BEH particles, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. They are designed for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features wide-pore (300Å) trifunctionally bonded BEH particles.
  • Offers wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Provides reproducible and consistent separations for proteomics, protein characterization, and peptide synthesis.
  • Produced in a cGMP, ISO 9002 accredited plant using ultra-pure reagents.
  • Each column is tested and includes a performance chromatogram and certificate.
Catalog No. 50-781-897