Learn More
Waters Corp Waters XBridge Peptide BEH C18 OBD Prep Column, 300Å, 5 μm, 19 mm × 100 mm
Catalog No. 50781907 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge BEH300 C18 Peptide Separation Technology (PST) columns are designed and quality control (QC) tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, offering a broad usable pH range, excellent low pH stability, and minimal column bleed for analyzing samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features a wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers a wide usable pH range (1 to 12) with superior low pH stability and ultra-low column bleed.
  • C18 chemistry for reversed phase separation.
  • Maximum pressure of 6000 psi (415 Bar).
  • Molecular weight range: 1,000 to 15,000.
  • Particle size: 5 μm; pore size: 300 Å.
  • 19 mm inner diameter, 100 mm length.
Catalog No. 50-781-907 Supplier Waters Corp Supplier No. 186003631
Add to Cart
Add to Cart
These XBridge BEH300 C18 Peptide Separation Technology (PST) columns are designed and quality control (QC) tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, offering a broad usable pH range, excellent low pH stability, and minimal column bleed for analyzing samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features a wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers a wide usable pH range (1 to 12) with superior low pH stability and ultra-low column bleed.
  • C18 chemistry for reversed phase separation.
  • Maximum pressure of 6000 psi (415 Bar).
  • Molecular weight range: 1,000 to 15,000.
  • Particle size: 5 μm; pore size: 300 Å.
  • 19 mm inner diameter, 100 mm length.