Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Peptide BEH C18 Column, 300Å, 5 μm, 1 mm x 50 mm

Catalog No. 50781891 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations.
  • Chemistry: C18.
  • Separation mode: Reversed phase.
  • Particle substrate: Hybrid.
  • pH range: 1 pH to 12 pH.
  • Maximum pressure: 6000 psi (415 bar).
  • Endcapped.
  • Low silanol activity.
  • Molecular weight range: 1000 to 15000.
  • Particle shape: Spherical.
  • Particle size: 5 μm.
  • Pore size: 300 Å.
  • System: HPLC.
  • Inner diameter: 1 mm.
  • Length: 50 mm.
  • Carbon load: 18%.

Catalog No. 50-781-891 Supplier Waters Corp Supplier No. 186003615
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations.
  • Chemistry: C18.
  • Separation mode: Reversed phase.
  • Particle substrate: Hybrid.
  • pH range: 1 pH to 12 pH.
  • Maximum pressure: 6000 psi (415 bar).
  • Endcapped.
  • Low silanol activity.
  • Molecular weight range: 1000 to 15000.
  • Particle shape: Spherical.
  • Particle size: 5 μm.
  • Pore size: 300 Å.
  • System: HPLC.
  • Inner diameter: 1 mm.
  • Length: 50 mm.
  • Carbon load: 18%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.