Learn More
Waters Corp Column, C18, 300Å, 5 μm, 2.1 mm x 150 mm
Supplier: Waters Corp 186003620
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
- Optimized and QC tested for peptide separations.
- Wide-pore (300Å) trifunctionally bonded BEH particle.
- Widest usable pH range, 1 pH to 12 pH.
- Superior low pH stability.
- Ultra-low column bleed.
- C18 chemistry.
- Maximum pressure of 6000 psi (415 Bar).
- Low silanol activity.
- Spherical particle shape with 5 μm particle size.
- Designed for HPLC systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.