Learn More
Waters Corp Waters Column, C4, 3.5 μm, 300 Å, 4.6 mm x 50 mm
Catalog No. 50782450 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is optimized for protein separation by size, hydrophobicity, and isoelectric point. Its C4 ligand minimizes protein carryover, increases protein recovery, and improves peak capacity. Using Peptide Separation Technology (PST), it is suitable for protein characterization and peptide synthesis.
  • QC tested and optimized for protein separation.
  • C4 ligand reduces protein carryover, boosts recovery, and improves peak capacity.
  • Wide-pore (300 Å) BEH particle for protein characterization and peptide synthesis.
  • Stable from pH 2 to 12 and up to 6,000 psi (415 Bar).
  • Packing materials developed in cGMP and ISO 9002 facility with ultra-pure reagents for peak shape, efficiency, and stability.
  • Each batch undergoes peptide separation tests for high-quality, repeatable separation.
Catalog No. 50-782-450 Supplier Waters Corp Supplier No. 186004502
This column is optimized for protein separation by size, hydrophobicity, and isoelectric point. Its C4 ligand minimizes protein carryover, increases protein recovery, and improves peak capacity. Using Peptide Separation Technology (PST), it is suitable for protein characterization and peptide synthesis.
  • QC tested and optimized for protein separation.
  • C4 ligand reduces protein carryover, boosts recovery, and improves peak capacity.
  • Wide-pore (300 Å) BEH particle for protein characterization and peptide synthesis.
  • Stable from pH 2 to 12 and up to 6,000 psi (415 Bar).
  • Packing materials developed in cGMP and ISO 9002 facility with ultra-pure reagents for peak shape, efficiency, and stability.
  • Each batch undergoes peptide separation tests for high-quality, repeatable separation.