Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Protein BEH C4 Column, 300 Å, 3.5 μm, 4.6 mm × 250 mm

Catalog No. 50782453 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This XBridge Protein BEH C4 Column is designed for protein separation in HPLC systems, offering lower retention compared to traditional C18 phases. It features a Hybrid particle substrate for reduced protein carryover, increased recovery, and improved peak capacity.

  • QC tested and optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • Features a spherical particle shape, low silanol activity, and Hybrid BEH particle technology.
  • Supports a pH range of 2 to 10 and withstands a maximum pressure of 6000 psi (415 Bar).
  • Particle size is 3.5 μm with a 300 Å pore size and 8% carbon load.
  • Internal dimensions are 4.6 mm diameter and 250 mm length.
  • Surface area is 90; molecular weight range maximum is 150000.
  • USP classification: L26.

Catalog No. 50-782-453 Supplier Waters Corp Supplier No. 186004505
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This XBridge Protein BEH C4 Column is designed for protein separation in HPLC systems, offering lower retention compared to traditional C18 phases. It features a Hybrid particle substrate for reduced protein carryover, increased recovery, and improved peak capacity.

  • QC tested and optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • Features a spherical particle shape, low silanol activity, and Hybrid BEH particle technology.
  • Supports a pH range of 2 to 10 and withstands a maximum pressure of 6000 psi (415 Bar).
  • Particle size is 3.5 μm with a 300 Å pore size and 8% carbon load.
  • Internal dimensions are 4.6 mm diameter and 250 mm length.
  • Surface area is 90; molecular weight range maximum is 150000.
  • USP classification: L26.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.