Learn More
Waters Corp WATERS CORPORATION XBridge Protein BEH C4 Column, 300Å, 3.5μm, 4.6 mm x 100 mm
Catalog No. 50782451 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH C4, 300Å packing material is engineered for protein separation, delivering excellent peak shape, high efficiency, and good recovery for challenging biological macromolecules.
  • Exceptional stability across a wide pH range (1 to 12) and high temperatures (20°C to 90°C).
  • Manufactured in a cGMP, ISO9002-certified plant using ultra-pure reagents, ensuring reproducible performance.
  • Provides excellent peak shape, high efficiency, and recovery for biological macromolecules.
  • Features a 300Å pore size.
  • Tolerates pressures up to 6000 psi (400 bar).
  • Individually tested for efficiency; includes a Performance Test Chromatogram and Certification of Acceptance.
  • eCord Intelligent Chip Technology provides a history of the column's performance (for XBridge XP 2.5 μm, <4.6 mm I.D. columns).
Catalog No. 50-782-451 Supplier Waters Corp Supplier No. 186004503
The XBridge Protein BEH C4, 300Å packing material is engineered for protein separation, delivering excellent peak shape, high efficiency, and good recovery for challenging biological macromolecules.
  • Exceptional stability across a wide pH range (1 to 12) and high temperatures (20°C to 90°C).
  • Manufactured in a cGMP, ISO9002-certified plant using ultra-pure reagents, ensuring reproducible performance.
  • Provides excellent peak shape, high efficiency, and recovery for biological macromolecules.
  • Features a 300Å pore size.
  • Tolerates pressures up to 6000 psi (400 bar).
  • Individually tested for efficiency; includes a Performance Test Chromatogram and Certification of Acceptance.
  • eCord Intelligent Chip Technology provides a history of the column's performance (for XBridge XP 2.5 μm, <4.6 mm I.D. columns).