Learn More
Waters Corp Waters XBridge Protein BEH C4 Column, 300 Å, 3.5 μm, 2.1 mm x 250 mm
Catalog No. 50782449 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH C4, 300 Å, 3.5 μm, 2.1 mm × 250 mm column is QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point. If you compare to traditional C18 phases, the C4 ligand is less retentive and will minimize protein carryover, increase protein recovery, and improve peak capacity.
  • QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point.
  • Less retentive C4 ligand compared to traditional C18 phases.
  • Minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.
Catalog No. 50-782-449 Supplier Waters Corp Supplier No. 186004501
The XBridge Protein BEH C4, 300 Å, 3.5 μm, 2.1 mm × 250 mm column is QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point. If you compare to traditional C18 phases, the C4 ligand is less retentive and will minimize protein carryover, increase protein recovery, and improve peak capacity.
  • QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point.
  • Less retentive C4 ligand compared to traditional C18 phases.
  • Minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.