Learn More
Waters Corp XBridge Protein BEH C4 Column, 300 Å, 3.5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186004504
Encompass_Preferred
This column is QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point. Compared to traditional C18 phases, the C4 ligand is less retentive, which minimizes protein carryover, increases protein recovery, and improves peak capacity. It provides excellent peak shape, high efficiency, and good recovery, making it suitable for biological macromolecules too large or hydrophobic for other columns.
  • Utilizes a C4 ligand for minimized protein carryover and improved peak capacity.
  • Offers superior stability across a wide pH range (2 to 10 pH) and at high temperatures.
  • Features a 300 Å pore size BEH particle (3.5 μm particle size) for ultra-low column bleed.
  • Manufactured under cGMP, ISO 9002 standards, with each column individually tested and certified.
Catalog No. 50-782-452