Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Prep Guard Cartridge, C18, 300Å, 10 μm, 10 mm X 10 mm

Catalog No. 50466008 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge BEH300 PREP C18, 10 μm, 10 x 10 mm Guard Column is designed to extend the life of preparative columns, featuring BEH300 PREP C18 packing material that meets peptide digest QC test criteria.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • QC tested: Peptide
  • Format: Prep guard cartridge
  • Surface area: 90
  • System: HPLC
  • Particle technology: BEH
  • USP classification: L1
  • Inner diameter: 10 mm
  • Length: 10 mm
  • Carbon load: 12%
  • Application: Peptide

Catalog No. 50-466-008 Supplier Waters Corp Supplier No. 186004467
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge BEH300 PREP C18, 10 μm, 10 x 10 mm Guard Column is designed to extend the life of preparative columns, featuring BEH300 PREP C18 packing material that meets peptide digest QC test criteria.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • QC tested: Peptide
  • Format: Prep guard cartridge
  • Surface area: 90
  • System: HPLC
  • Particle technology: BEH
  • USP classification: L1
  • Inner diameter: 10 mm
  • Length: 10 mm
  • Carbon load: 12%
  • Application: Peptide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.