Learn More
Waters Corp Waters Prep Guard Cartridge, C18, 10 μm, 300 Å
Supplier:  Waters Corp 186004466
Encompass_Preferred
This preparative guard cartridge is designed to extend the life of preparative columns. It features BEH C18 chemistry, a 300 Å pore size, and a 10 μm particle size. With dimensions of 19 mm inner diameter x 10 mm length, it is compatible with HPLC systems and suitable for peptide applications, offering a pH range of 1 to 12.
  • Designed to extend the life of preparative columns.
  • Features BEH C18 chemistry.
  • 300 Å pore size and 10 μm particle size.
  • Compatible with HPLC systems.
  • Suitable for peptide applications.
  • pH range of 1 to 12.
Catalog No. 50-466-007