Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Prep Guard Cartridge, C18, 300Å, 5 μm

Catalog No. 50466011 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Prep Guard Cartridge was developed to extend the life of the BEH300, C18, 10 μm, 10mm ID preparative column. The packing material used must pass the peptide digest, QC test criteria established for the prep column.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • PH range: 1 pH to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 300 Å
  • QC tested: Peptide
  • Surface area: 90
  • System: HPLC
  • Particle technology: BEH
  • USP classification: L1
  • Inner diameter: 10 mm
  • Length: 10 mm
  • Carbon load: 12 %
  • Application: Peptide
  • Units per package: 1 pk

Catalog No. 50-466-011 Supplier Waters Corp Supplier No. 186004471
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This Prep Guard Cartridge was developed to extend the life of the BEH300, C18, 10 μm, 10mm ID preparative column. The packing material used must pass the peptide digest, QC test criteria established for the prep column.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • PH range: 1 pH to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 300 Å
  • QC tested: Peptide
  • Surface area: 90
  • System: HPLC
  • Particle technology: BEH
  • USP classification: L1
  • Inner diameter: 10 mm
  • Length: 10 mm
  • Carbon load: 12 %
  • Application: Peptide
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.