Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Prep Guard Cartridge, C18, 5 μm, 300 Å

Catalog No. 50466010 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This guard cartridge is designed for preparative HPLC systems. It provides robust protection for preparative columns, extending their lifespan and ensuring consistent performance in peptide applications.

  • C18 chemistry for reversed-phase separation
  • Hybrid particle substrate
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped to reduce silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Pore size: 300 Å
  • QC tested for peptide applications
  • Format: Prep Guard Cartridge
  • Compatible with HPLC systems
  • BEH particle technology
  • USP Classification: L1
  • Inner diameter: 19 mm
  • Length: 10 mm
  • Carbon load: 12%

Catalog No. 50-466-010 Supplier Waters Corp Supplier No. 186004470
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This guard cartridge is designed for preparative HPLC systems. It provides robust protection for preparative columns, extending their lifespan and ensuring consistent performance in peptide applications.

  • C18 chemistry for reversed-phase separation
  • Hybrid particle substrate
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped to reduce silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Pore size: 300 Å
  • QC tested for peptide applications
  • Format: Prep Guard Cartridge
  • Compatible with HPLC systems
  • BEH particle technology
  • USP Classification: L1
  • Inner diameter: 19 mm
  • Length: 10 mm
  • Carbon load: 12%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.