Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 300Å, 10 μm, 4.6 mm x 50 mm
Catalog No. 50781935 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are specifically designed and quality control tested for peptide separations. These columns feature wide-pore (300Å) trifunctionally bonded BEH particles, offering a broad usable pH range, excellent stability at low pH, and very low column bleed. This makes them ideal for analyzing samples in proteomics, protein characterization, and peptide synthesis applications.
  • Optimized and QC tested for peptide separations
  • Wide-pore (300Å) trifunctionally bonded BEH particle
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed
Catalog No. 50-781-935 Supplier Waters Corp Supplier No. 186003663
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are specifically designed and quality control tested for peptide separations. These columns feature wide-pore (300Å) trifunctionally bonded BEH particles, offering a broad usable pH range, excellent stability at low pH, and very low column bleed. This makes them ideal for analyzing samples in proteomics, protein characterization, and peptide synthesis applications.
  • Optimized and QC tested for peptide separations
  • Wide-pore (300Å) trifunctionally bonded BEH particle
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed