Learn More
Waters Corp Waters XBridge Peptide BEH C18 OBD Prep Column, 300 Å, 10 μm, 30 mm x 50 mm
Catalog No. 50781946 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH300 C18 Peptide Separation Technology (PST) preparative columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Optimized and QC tested for peptide separations
  • Features a wide-pore (300Å) trifunctionally bonded BEH particle
  • Offers the widest usable pH range
  • Provides superior low pH stability
  • Ensures ultra-low column bleed
  • Suitable for proteomics, protein characterization, and peptide synthesis applications
  • Utilizes Optimum Bed Density (OBD) hardware format
  • Ensures superior column lifetime
Catalog No. 50-781-946 Supplier Waters Corp Supplier No. 186003674
Add to Cart
Add to Cart
Request a Quote
These BEH300 C18 Peptide Separation Technology (PST) preparative columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Optimized and QC tested for peptide separations
  • Features a wide-pore (300Å) trifunctionally bonded BEH particle
  • Offers the widest usable pH range
  • Provides superior low pH stability
  • Ensures ultra-low column bleed
  • Suitable for proteomics, protein characterization, and peptide synthesis applications
  • Utilizes Optimum Bed Density (OBD) hardware format
  • Ensures superior column lifetime