Learn More
Waters Corp Waters Corporation XBridge Peptide BEH C18 OBD Column, 19 x 50 mm, 10 μm Particle, 300 Å Pore
Catalog No. 50781943 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18, 300 Å OBD Column is a high-performance column designed for preparative applications. It features specific QC testing to ensure consistent column-to-column performance, making it suitable for a wide range of peptide and small protein separations.
  • Features trifunctional C18 ligand, fully end-capped, bonded to Ethylene-Bridged Hybrid (BEH) particles.
  • Offers outstanding peak capacity and superior peak shape in trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents.
  • Provides different separation selectivities for a wide range of peptides and small proteins with a 300 Å pore size.
  • Each synthesized batch is specifically QC tested with a complex protein digest to help ensure reproducible preparative column performance.
Catalog No. 50-781-943 Supplier Waters Corp Supplier No. 186003671
The XBridge Peptide BEH C18, 300 Å OBD Column is a high-performance column designed for preparative applications. It features specific QC testing to ensure consistent column-to-column performance, making it suitable for a wide range of peptide and small protein separations.
  • Features trifunctional C18 ligand, fully end-capped, bonded to Ethylene-Bridged Hybrid (BEH) particles.
  • Offers outstanding peak capacity and superior peak shape in trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents.
  • Provides different separation selectivities for a wide range of peptides and small proteins with a 300 Å pore size.
  • Each synthesized batch is specifically QC tested with a complex protein digest to help ensure reproducible preparative column performance.