Learn More
Waters Corp Waters XBridge Column, Peptide BEH C18, 300Å, 10 μm
Supplier:  Waters Corp 186003673
Encompass_Preferred
This preparative column is optimized and QC tested for peptide separations. It features a wide-pore (300Å) trifunctionally bonded BEH particle, offering a wide usable pH range and superior low pH stability. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under various operating conditions.
  • Optimized and QC tested for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
  • Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
Catalog No. 50-781-945
Add to Cart
Request a Quote