Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 300 Å, 30 × 100 mm, 10 μm
Supplier:  Waters Corp 186003675
Encompass_Preferred
Waters OBD Preparative Columns are engineered for preparative liquid chromatography, providing direct scalability from analytical to preparative LC, exceptional column lifetime, and consistent performance. This XBridge Peptide BEH C18, 300 Å column ensures excellent resolution, peak shape, and mass loading for isolating targeted compounds.
  • Uniform, optimally packed column beds deliver ideal outcomes in preparative chromatography.
  • Exceptional column lifetimes ensure long-lasting performance.
  • Uncompromised scale-up allows seamless transition from analytical to preparative chromatography.
  • Maximized recoveries are achieved with robust packings designed for isolating targeted small molecules and biomolecules.
  • Decreased risk reduces the need for additional Prep method development.
Catalog No. 50-781-947
Add to Cart
Request a Quote