Learn More
Waters Corp XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 µm, 4.6 mm X 150 mm, 1/pk
Catalog No. NC2331035 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 µm, 4.6 mm X 150 mm, 1/pk
Catalog No. NC2331035 Supplier Waters Corp Supplier No. 186010547
XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 µm, 4.6 mm X 150 mm, 1/pk