Learn More
Waters Corp Waters XBridge BEH C18 OBD Prep Column, 130Å, 5 μm, 50 mm × 150 mm
Catalog No. 50596828 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This OBD Prep Column is engineered for fast, efficient, lab-scale separations, offering high productivity and extended column lifetimes. Its trifunctionally bonded C18 ligand provides a universal solution for most HPLC separations across a wide pH range, with superior low pH stability and ultra-low column bleed, while OBD technology ensures exceptional resistance to mechanical bed failure.
  • Designed for fast, efficient, lab-scale separations.
  • Offers higher recoveries and extended column lifetimes.
  • Enables direct scaling from UPLC, UHPLC, or HPLC to lab-scale purification.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Provides widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • OBD technology ensures exceptional resistance to mechanical bed failure.
Catalog No. 50-596-828 Supplier Waters Corp Supplier No. 186003929
Add to Cart
Add to Cart
Request a Quote
This OBD Prep Column is engineered for fast, efficient, lab-scale separations, offering high productivity and extended column lifetimes. Its trifunctionally bonded C18 ligand provides a universal solution for most HPLC separations across a wide pH range, with superior low pH stability and ultra-low column bleed, while OBD technology ensures exceptional resistance to mechanical bed failure.
  • Designed for fast, efficient, lab-scale separations.
  • Offers higher recoveries and extended column lifetimes.
  • Enables direct scaling from UPLC, UHPLC, or HPLC to lab-scale purification.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Provides widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • OBD technology ensures exceptional resistance to mechanical bed failure.