Learn More
Waters Corp WATERS CORPORATION Column, C18, 130Å, 3.5 μm, 1 mm x 50 mm
Catalog No. 50781562 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is a workhorse for HPLC method development, offering a universal column for most HPLC separations. It provides the widest usable pH range (1–12), superior low pH stability, and ultra-low column bleed. Designed to increase productivity, it is suitable for quality control and cutting-edge LC/MS assays.
  • Outstanding peak shape and capacity
  • Reduced retention loss and peak shape deterioration
  • Proprietary bonding and end-capping ensures ligand stability and chromatographic repeatability at low pH
  • Consistent performance throughout its lifespan
  • C18 chemistry and hybrid particle substrate for reversed phase separation
  • Maximum pressure: 6000 psi (415 Bar)
Catalog No. 50-781-562 Supplier Waters Corp Supplier No. 186003126
Add to Cart
Add to Cart
The XBridge BEH C18 sorbent is a workhorse for HPLC method development, offering a universal column for most HPLC separations. It provides the widest usable pH range (1–12), superior low pH stability, and ultra-low column bleed. Designed to increase productivity, it is suitable for quality control and cutting-edge LC/MS assays.
  • Outstanding peak shape and capacity
  • Reduced retention loss and peak shape deterioration
  • Proprietary bonding and end-capping ensures ligand stability and chromatographic repeatability at low pH
  • Consistent performance throughout its lifespan
  • C18 chemistry and hybrid particle substrate for reversed phase separation
  • Maximum pressure: 6000 psi (415 Bar)