Learn More
Waters Corp Waters XBridge BEH C18 Column, 130Å, 3.5 μm, 2.1 mm × 50 mm
Catalog No. 50781474 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is a universal column for HPLC method development, usable across a mobile phase pH range of 1–12. It provides superior low pH stability, ultra–low column bleed, and ensures reliable performance through controlled manufacturing. Designed for versatility, it supports LC/MS assays and quality control methods, offering increased column lifetime, enhanced reliability, and unmatched peak shape.
  • Can be used over an entire range of mobile phase pH (1–12)
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations
  • Ensures product reproducibility through testing from raw material characterization to final batch analysis
  • Designed for unrivaled versatility, suitable for developing LC/MS assays or quality control methods
Catalog No. 50-781-474 Supplier Waters Corp Supplier No. 186003021
Add to Cart
Add to Cart
The XBridge BEH C18 sorbent is a universal column for HPLC method development, usable across a mobile phase pH range of 1–12. It provides superior low pH stability, ultra–low column bleed, and ensures reliable performance through controlled manufacturing. Designed for versatility, it supports LC/MS assays and quality control methods, offering increased column lifetime, enhanced reliability, and unmatched peak shape.
  • Can be used over an entire range of mobile phase pH (1–12)
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations
  • Ensures product reproducibility through testing from raw material characterization to final batch analysis
  • Designed for unrivaled versatility, suitable for developing LC/MS assays or quality control methods