Learn More
Waters Corp XBridge BEH C18 Column, 130Å, 3.5 µm, 2.1 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003023
Encompass_Preferred
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Catalog No. 50-781-476