Learn More
Waters Corp Waters Column, C18, 130Å, 3.5 μm, 3 mm × 150 mm
Catalog No. 50781481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development, offering compatibility across a mobile phase pH range of 1-12. This column features a trifunctionally bonded C18 ligand, providing universal utility for most HPLC separations, with superior low pH stability and ultra-low column bleed.
  • Compatible with mobile phase pH from 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Provides superior low pH stability and ultra-low column bleed.
  • Allows operation under various mobile phase, temperature, and pH conditions.
  • Offers maximum stability under extreme pH conditions.
  • Minimizes LC/MS bleed and enhances hydrolytic stability and particle strength.
  • Optimized for control over analyte retention and selectivity.
Catalog No. 50-781-481 Supplier Waters Corp Supplier No. 186003028
Add to Cart
Add to Cart
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development, offering compatibility across a mobile phase pH range of 1-12. This column features a trifunctionally bonded C18 ligand, providing universal utility for most HPLC separations, with superior low pH stability and ultra-low column bleed.
  • Compatible with mobile phase pH from 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Provides superior low pH stability and ultra-low column bleed.
  • Allows operation under various mobile phase, temperature, and pH conditions.
  • Offers maximum stability under extreme pH conditions.
  • Minimizes LC/MS bleed and enhances hydrolytic stability and particle strength.
  • Optimized for control over analyte retention and selectivity.