Learn More
Waters Corp XBridge C18 Column, 3.5 μm, 3.0 x 30 mm
Supplier:  Waters Corp 186003025
Encompass_Preferred
The XBridge C18 Column utilizes patented BEH Technology, offering high chemical and mechanical stability across a wide pH range of 1 to 12. Designed for exceptional performance and longevity, this column provides reliable and reproducible results for various chromatographic applications.
  • Utilizes BEH Technology for high chemical and mechanical stability.
  • Offers exceptional pH stability, operating within a pH range of 1 to 12.
  • Provides increased column lifetime due to resistance to particle dissolution.
  • Reduces surface silanol concentration for improved peak shape and lower USP peak tailing factors.
  • Enables method scalability and seamless migration across UPLC, analytical, and preparative platforms.
  • Ensures reliable and reproducible results under harsh operating conditions.
  • Virtually eliminates LC/MS bleed due to increased hydrolytic stability and particle strength.
Catalog No. 50-781-478