Learn More
Waters Corp Waters XBridge BEH C18 Column, 130Å, 3.5 μm, 4.6 mm x 100 mm
Supplier:  Waters Corp 186003033
Encompass_Preferred
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development, capable of use across an entire mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand provides a universal column suitable for most HPLC separations, offering the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Achieves superior low and high pH stability, leading to outstanding chromatographic peaks.
  • Features a trifunctional bonding and end-capping process that overcomes low-pH instability from siloxane bond cleavage and reduces hydrolysis.
  • Incorporates ethylene bridges within the silica matrix to resist particle dissolution, thereby preventing column voids, efficiency loss, and peak degradation at elevated pH.
  • Offers unrivaled flexibility for methods development.
Catalog No. 50-781-486