Learn More
Waters Corp XBridge Column, C18, 130 Å, 3.5 μm
Supplier:  Waters Corp 186003943
Encompass_Preferred
The XBridge BEH C18 Column is designed for HPLC method development, offering a universal solution with its trifunctionally bonded C18 ligand. It provides a wide mobile phase pH range of 1 to 12, superior low pH stability, and ultra-low column bleed, ensuring consistently accurate results for rigorous assays.
  • Universal column for HPLC separations with trifunctionally bonded C18 ligand.
  • Widest usable pH range of 1 to 12.
  • Superior low pH stability and ultra-low column bleed.
  • Maximizes particle efficiency and provides unmatched peak shape and peak capacity.
  • Enhanced hydrolytic stability and particle strength due to BEH technology, minimizing LC/MS bleed.
  • Ensures consistent performance through tightly controlled manufacturing and extensive testing.
Catalog No. 50-782-077