Learn More
Waters Corp Waters XBridge BEH C18 Column, 130Å, 3.5 μm, 4.6 mm × 30 mm
Catalog No. 50781483 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC column is designed for robust performance across a wide pH range of 1-12. Featuring BEH C18 sorbent and a trifunctionally bonded C18 ligand, it offers superior low pH stability and ultra-low column bleed, suitable for various HPLC and LC/MS applications. It ensures enhanced pH stability for longer column life and improved assay dependability.
  • Utilizes BEH C18 sorbent across a mobile phase pH range of 1-12 for HPLC method development.
  • Offers a universal column for most HPLC separations with trifunctionally bonded C18 ligand.
  • Provides superior stability at low pH and ultra-low column bleed for robust assays.
  • Designed for extended column lifetimes and improved column dependability.
  • Achieves exceptional peak shape and peak capacity through increased particle efficiency.
  • Seamless transition with UltraPerformance LC technology.
Catalog No. 50-781-483 Supplier Waters Corp Supplier No. 186003030
This HPLC column is designed for robust performance across a wide pH range of 1-12. Featuring BEH C18 sorbent and a trifunctionally bonded C18 ligand, it offers superior low pH stability and ultra-low column bleed, suitable for various HPLC and LC/MS applications. It ensures enhanced pH stability for longer column life and improved assay dependability.
  • Utilizes BEH C18 sorbent across a mobile phase pH range of 1-12 for HPLC method development.
  • Offers a universal column for most HPLC separations with trifunctionally bonded C18 ligand.
  • Provides superior stability at low pH and ultra-low column bleed for robust assays.
  • Designed for extended column lifetimes and improved column dependability.
  • Achieves exceptional peak shape and peak capacity through increased particle efficiency.
  • Seamless transition with UltraPerformance LC technology.