Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge BEH C18 Column, 130Å, 3.5 μm, 4.6 mm X 50 mm

Catalog No. 50781484 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge BEH C18 sorbent is designed for HPLC method development and can be utilized across a mobile phase pH range of 1–12. Featuring a trifunctionally bonded C18 ligand, it offers a wide usable pH range, superior low pH stability, and ultra-low column bleed for most HPLC separations.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle size: 3.5 μm
  • Pore size: 130 Å
  • Dimensions: 4.6 mm X 50 mm
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 Bar)
  • BEH particle technology
  • Excellent reproducibility and column lifetime
  • Compatible with phosphate buffers

Catalog No. 50-781-484 Supplier Waters Corp Supplier No. 186003031
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge BEH C18 sorbent is designed for HPLC method development and can be utilized across a mobile phase pH range of 1–12. Featuring a trifunctionally bonded C18 ligand, it offers a wide usable pH range, superior low pH stability, and ultra-low column bleed for most HPLC separations.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle size: 3.5 μm
  • Pore size: 130 Å
  • Dimensions: 4.6 mm X 50 mm
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 Bar)
  • BEH particle technology
  • Excellent reproducibility and column lifetime
  • Compatible with phosphate buffers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.