Learn More
Waters Corp Waters XBridge BEH C18 Column, 130Å, 5 μm, 2.1 mm x 150 mm
Catalog No. 50781550 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BEH C18 Column is designed for HPLC method development, offering a broad mobile phase pH range (1 to 12). It features a trifunctionally bonded C18 ligand, providing a universal column for most HPLC separations with superior low pH stability and ultra-low column bleed. This column delivers high separation performance, resilience, and throughput for HPLC tests, facilitating method transfer between HPLC and UPLC.
  • Universal column for most HPLC separations with a mobile phase pH range of 1 to 12.
  • Superior low pH stability and ultra-low column bleed.
  • Delivers high separation performance, resilience, and throughput for HPLC tests.
  • Designed to tolerate higher pressures and ensures unrivaled column longevity.
  • Simplifies technique transfer between HPLC and UPLC.
Catalog No. 50-781-550 Supplier Waters Corp Supplier No. 186003110
The BEH C18 Column is designed for HPLC method development, offering a broad mobile phase pH range (1 to 12). It features a trifunctionally bonded C18 ligand, providing a universal column for most HPLC separations with superior low pH stability and ultra-low column bleed. This column delivers high separation performance, resilience, and throughput for HPLC tests, facilitating method transfer between HPLC and UPLC.
  • Universal column for most HPLC separations with a mobile phase pH range of 1 to 12.
  • Superior low pH stability and ultra-low column bleed.
  • Delivers high separation performance, resilience, and throughput for HPLC tests.
  • Designed to tolerate higher pressures and ensures unrivaled column longevity.
  • Simplifies technique transfer between HPLC and UPLC.