Learn More
Waters Corp Waters XBridge BEH C18 Column, 5 μm, 2.1 mm, 30 mm
Catalog No. 50781565 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BEH C18 sorbent is engineered for high-performance liquid chromatography (HPLC) method development, functioning across a wide mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column for diverse HPLC separations, providing superior low pH stability and ultra-low column bleed, ideal for LC/MS assays or quality control.
  • Functions across a mobile phase pH range of 1-12.
  • Offers superior low pH stability and ultra-low column bleed.
  • Extends column lifetime and reduces lab costs via enhanced pH stability.
  • Improves column reliability and assay robustness.
  • Delivers exceptional peak shape and capacity by increasing particle efficiency.
  • Utilizes specialized bonding and end-capping for ligand stability at low pH.
  • Exhibits minimal retention loss or peak shape deterioration.
  • Demonstrates high durability under high pH settings.
Catalog No. 50-781-565 Supplier Waters Corp Supplier No. 186003129
This BEH C18 sorbent is engineered for high-performance liquid chromatography (HPLC) method development, functioning across a wide mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column for diverse HPLC separations, providing superior low pH stability and ultra-low column bleed, ideal for LC/MS assays or quality control.
  • Functions across a mobile phase pH range of 1-12.
  • Offers superior low pH stability and ultra-low column bleed.
  • Extends column lifetime and reduces lab costs via enhanced pH stability.
  • Improves column reliability and assay robustness.
  • Delivers exceptional peak shape and capacity by increasing particle efficiency.
  • Utilizes specialized bonding and end-capping for ligand stability at low pH.
  • Exhibits minimal retention loss or peak shape deterioration.
  • Demonstrates high durability under high pH settings.