Learn More
Waters Corp Waters Column, C18, 5 μm, 130 Å, 3 mm x 30 mm
Catalog No. 50781551 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BEH C18 column can be used over an entire range of mobile phase pH (1-12). Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Can be used over an entire range of mobile phase pH (1-12).
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations.
  • Offers the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Operates optimally within 60 – 80 °C range; however, operating close to or outside limits can impact column lifetime and performance.
Catalog No. 50-781-551 Supplier Waters Corp Supplier No. 186003111
This BEH C18 column can be used over an entire range of mobile phase pH (1-12). Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Can be used over an entire range of mobile phase pH (1-12).
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations.
  • Offers the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Operates optimally within 60 – 80 °C range; however, operating close to or outside limits can impact column lifetime and performance.