Learn More
Waters Corp Waters XBridge BEH C18 Column, 130Å, 5 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186003131
Encompass_Preferred
The XBridge BEH C18 Column is designed for HPLC method development, offering a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand provides superior low pH stability and ultra-low column bleed. BEH Technology enhances hydrolytic stability and particle strength, preventing LC/MS bleed and ensuring exceptional separation performance.
  • Increased hydrolytic stability and particle strength via BEH Technology.
  • Eliminates LC/MS bleed.
  • Exceptional separation performance and throughput.
  • Wide usable pH range (1-12) with C18 ligand.
  • Superior low pH stability and ultra-low column bleed.
  • Maintains retention and peak shape at low pH.
  • Improves peak shape via ethylene bridge technology.
  • Optimal performance at 60–80°C.
Catalog No. 50-781-567