Learn More
Waters Corp Waters Column, C18, 5 μm, 3 mm, 100 mm
Catalog No. 50781568 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development and is suitable for use across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Utilizes patented trifunctional bonding and end-capping technology to prevent low-pH instability caused by siloxane bond cleavage via acid hydrolysis.
  • Ethylene bridges within the silica matrix address issues associated with silica-based chromatographic particle failure at high pH, such as particle dissolving, column voids, efficiency loss, and peak degradation.
  • Produced in cGMP, ISO9001, and ISO13485 certified facilities, ensuring exceptional reproducibility and consistent column performance.
Catalog No. 50-781-568 Supplier Waters Corp Supplier No. 186003132
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development and is suitable for use across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Utilizes patented trifunctional bonding and end-capping technology to prevent low-pH instability caused by siloxane bond cleavage via acid hydrolysis.
  • Ethylene bridges within the silica matrix address issues associated with silica-based chromatographic particle failure at high pH, such as particle dissolving, column voids, efficiency loss, and peak degradation.
  • Produced in cGMP, ISO9001, and ISO13485 certified facilities, ensuring exceptional reproducibility and consistent column performance.