Learn More
Waters Corp Waters Corporation XBridge BEH C18 Column, 5 μm, 4.6 mm, 150 mm
Catalog No. 50781556 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 Column is a robust and versatile tool for HPLC method development and routine separations. It features trifunctionally bonded C18 ligands on ethylene bridged hybrid (BEH) particles, providing excellent chemical stability across a wide pH range (1-12). This column is designed to offer superior low pH stability and ultra-low column bleed, ensuring reproducible, high-quality data for critical applications in various analytical fields.
  • Usable over an entire range of mobile phase pH (1-12).
  • Trifunctionally bonded C18 ligand.
  • Provides the widest usable pH range.
  • Offers superior low pH stability.
  • Exhibits ultra-low column bleed.
  • Ensures reproducible, high-quality data.
  • Suitable for pharmaceutical, environmental, and biochemical analysis.
Catalog No. 50-781-556 Supplier Waters Corp Supplier No. 186003116
Add to Cart
Add to Cart
The XBridge BEH C18 Column is a robust and versatile tool for HPLC method development and routine separations. It features trifunctionally bonded C18 ligands on ethylene bridged hybrid (BEH) particles, providing excellent chemical stability across a wide pH range (1-12). This column is designed to offer superior low pH stability and ultra-low column bleed, ensuring reproducible, high-quality data for critical applications in various analytical fields.
  • Usable over an entire range of mobile phase pH (1-12).
  • Trifunctionally bonded C18 ligand.
  • Provides the widest usable pH range.
  • Offers superior low pH stability.
  • Exhibits ultra-low column bleed.
  • Ensures reproducible, high-quality data.
  • Suitable for pharmaceutical, environmental, and biochemical analysis.