Learn More
Waters Corp Waters Corporation Column, XBridge BEH C18, 5 μm, 4.6 mm x 100 mm
Supplier:  Waters Corp 186003115
Encompass_Preferred
This C18 sorbent is designed for high-performance liquid chromatography (HPLC) method development. It offers a versatile solution for a wide range of separations, featuring a trifunctionally bonded C18 ligand that provides superior stability across a mobile phase pH range of 1 to 12, excellent low pH stability, and ultra-low column bleed.
  • Designed for HPLC method development, usable across a mobile phase pH range of 1 to 12.
  • Features a trifunctionally bonded C18 ligand for universal HPLC separations.
  • Offers superior low pH stability and ultra-low column bleed for reliable results.
  • Ensures excellent reproducibility, performance, and extended column lifetime.
  • Utilizes BEH Technology particles for stable operation at higher pH conditions and seamless pH switching.
  • Includes documentation, a certificate of analysis, and a performance test chromatogram.
Catalog No. 50-781-555