Learn More
Waters Corp Waters XBridge BEH C18 Column, 5 μm, 4.6 mm X 250 mm, 130 Å
Supplier:  Waters Corp 186003117
Encompass_Preferred
The XBridge BEH C18 Column is a high-performance liquid chromatography (HPLC) column designed for robust and reproducible separations. It features a trifunctionally bonded C18 ligand and hybrid particle substrate, offering broad pH stability (1 to 12) and superior low pH stability.
  • Can be used over an entire range of mobile phase pH (1 to 12).
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations.
  • Offers the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Unparalleled column lifespan.
  • Allows seamless migration to UPLC technology where needed.
  • BEH technology enables the full power of pH to be used in routine methods development.
  • Delivers excellent peak shape, high efficiency, and excellent stability for acidic and basic mobile phases.
Catalog No. 50-781-557