Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Method Validation Kit, C18, 3.5 µm, 130 Å, 4.6 mm x 150 mm

Catalog No. 50782025 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Method Validation Kit utilizes C18 sorbent for HPLC method development, accommodating a mobile phase pH range of 1–12. This analytical column is designed to maximize productivity for LC/MS assays and quality control methods, offering superior low pH stability for increased column lifetime and enhanced assay ruggedness.

  • Accommodates mobile phase pH range of 1–12.
  • Trifunctionally bonded C18 ligand.
  • Superior low pH stability and ultra-low column bleed.
  • Increased column lifetime and enhanced assay ruggedness.
  • Maximizes particle efficiency for unmatched peak shape and peak capacity.
  • Reproducible column performance through tightly controlled manufacturing.
  • Particle size: 3.5 µm.
  • Pore size: 130 Å.
  • Inner diameter: 4.6 mm.
  • Length: 150 mm.
  • Units per package: 3.

Catalog No. 50-782-025 Supplier Waters Corp Supplier No. 186003770
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Method Validation Kit utilizes C18 sorbent for HPLC method development, accommodating a mobile phase pH range of 1–12. This analytical column is designed to maximize productivity for LC/MS assays and quality control methods, offering superior low pH stability for increased column lifetime and enhanced assay ruggedness.

  • Accommodates mobile phase pH range of 1–12.
  • Trifunctionally bonded C18 ligand.
  • Superior low pH stability and ultra-low column bleed.
  • Increased column lifetime and enhanced assay ruggedness.
  • Maximizes particle efficiency for unmatched peak shape and peak capacity.
  • Reproducible column performance through tightly controlled manufacturing.
  • Particle size: 3.5 µm.
  • Pore size: 130 Å.
  • Inner diameter: 4.6 mm.
  • Length: 150 mm.
  • Units per package: 3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.