Learn More
Waters Corp Waters Corporation Column Kit, XBridge BEH C18, 3.5 μm, 3 mm x 100 mm
Catalog No. 50782022 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development and can be utilized across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped for enhanced performance
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 130 Å pore size
  • Method validation kit format
  • 185 surface area
  • Designed for HPLC system
  • BEH particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 100 mm length
  • 18 % carbon load
  • 3 units per package
Catalog No. 50-782-022 Supplier Waters Corp Supplier No. 186003767
Add to Cart
Add to Cart
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development and can be utilized across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped for enhanced performance
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 130 Å pore size
  • Method validation kit format
  • 185 surface area
  • Designed for HPLC system
  • BEH particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 100 mm length
  • 18 % carbon load
  • 3 units per package