Learn More
Waters Corp XBridge BEH C18 Method Validation Kit, 130Å, 5 µm, 4.6 mm X 100 mm, 3/pk
Catalog No. 50782029 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Catalog No. 50-782-029 Supplier Waters Corp Supplier No. 186003774
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.