Learn More
Waters Corp Waters Corporation Sentry Guard Cartridge, C18, 3.5 μm, 3 mm x 20 mm, 130 Å
Catalog No. 50791144 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge protects analytical columns from particle and chemical contamination, extending life and improving performance. Its BEH C18 sorbent operates across a pH range of 1 to 12, offering universal HPLC separation with high stability. Compatible with 3 mm I.D. BEH C18 analytical columns (3.5 μm particle size), it withstands pressures up to 6,000 psi (415 bar) and is Q.C. tested.
  • Protects columns from contamination.
  • Extends column life; improves performance.
  • Wide pH range: 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC.
  • High stability, ultra-low column bleed.
  • Easy setup for UPLC columns.
  • Withstands 6,000 psi (415 bar).
  • Q.C. tested for consistent performance.
Catalog No. 50-791-144 Supplier Waters Corp Supplier No. 186003060
This Sentry Guard Cartridge protects analytical columns from particle and chemical contamination, extending life and improving performance. Its BEH C18 sorbent operates across a pH range of 1 to 12, offering universal HPLC separation with high stability. Compatible with 3 mm I.D. BEH C18 analytical columns (3.5 μm particle size), it withstands pressures up to 6,000 psi (415 bar) and is Q.C. tested.
  • Protects columns from contamination.
  • Extends column life; improves performance.
  • Wide pH range: 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC.
  • High stability, ultra-low column bleed.
  • Easy setup for UPLC columns.
  • Withstands 6,000 psi (415 bar).
  • Q.C. tested for consistent performance.