Learn More
Waters Corp Waters Corporation Sentry Guard Cartridge, 130Å, 5 μm, 2.1 x 10 mm
Catalog No. 50781514 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is designed for HPLC method development, offering utility across a wide range of mobile phase pH from 1 to 12. Its trifunctionally bonded C18 ligand ensures universal application for most HPLC separations, providing a broad usable pH range, excellent low pH stability, and minimal column bleed.
  • Increases the lifespan of the XBridge BEH C18 Column.
  • Limits the entry of contaminating chemicals and suspended particles into the analytical column.
  • Made of high-performance packing materials used in analytical columns.
  • Specifically designed for situations requiring a higher level of biological fluid filtration and/or temperature control.
Catalog No. 50-781-514 Supplier Waters Corp Supplier No. 186003062
The XBridge BEH C18 sorbent is designed for HPLC method development, offering utility across a wide range of mobile phase pH from 1 to 12. Its trifunctionally bonded C18 ligand ensures universal application for most HPLC separations, providing a broad usable pH range, excellent low pH stability, and minimal column bleed.
  • Increases the lifespan of the XBridge BEH C18 Column.
  • Limits the entry of contaminating chemicals and suspended particles into the analytical column.
  • Made of high-performance packing materials used in analytical columns.
  • Specifically designed for situations requiring a higher level of biological fluid filtration and/or temperature control.