Learn More
Waters Corp HPLC Column, XBridge BEH C18, 3.5 μm, 130 Å, 3 x 20 mm
Catalog No. 50781477 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-performance column is ideal for HPLC method development across a mobile phase pH range of 1 to 12. Its trifunctionally bound C18 ligand provides universal applicability for most HPLC separations, offering a wide usable pH range, excellent low pH stability, and minimal column bleed. Utilizing Ethylene Bridge Hybrid Particle technology, it withstands high pH, high temperatures, and extreme operating conditions, making it suitable for a wide range of LC-MS analyses of small molecules and biomolecules.
  • High-performance column for HPLC method development.
  • Usable over a pH range of 1 to 12.
  • Provides excellent stability at low pH values.
  • Ensures extremely low column bleed.
  • Withstands high pH, high temperatures, and extreme operating conditions.
  • Suitable for a wide range of LC-MS analyses.
Catalog No. 50-781-477 Supplier Waters Corp Supplier No. 186003024
This high-performance column is ideal for HPLC method development across a mobile phase pH range of 1 to 12. Its trifunctionally bound C18 ligand provides universal applicability for most HPLC separations, offering a wide usable pH range, excellent low pH stability, and minimal column bleed. Utilizing Ethylene Bridge Hybrid Particle technology, it withstands high pH, high temperatures, and extreme operating conditions, making it suitable for a wide range of LC-MS analyses of small molecules and biomolecules.
  • High-performance column for HPLC method development.
  • Usable over a pH range of 1 to 12.
  • Provides excellent stability at low pH values.
  • Ensures extremely low column bleed.
  • Withstands high pH, high temperatures, and extreme operating conditions.
  • Suitable for a wide range of LC-MS analyses.